Learn More
Sino Biological Recombinant Cynomolgus Siglec-15 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 90930C02H200
Encompass_Preferred
A DNA sequence encoding the cynomolgus SIGLEC15 (A0A2K5UY47) (Met1-Thr263) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2648