Learn More
Sino Biological Recombinant Cynomolgus Siglec-6 Protein (ECD, His Tag)
Catalog No. 502992741
Encompass_Preferred
A DNA sequence encoding the Cynomolgus SIGLEC6 (XP_005590181.3) (Met1-Gly333) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2741 Supplier Sino Biological Supplier No. 91063C08H1
A DNA sequence encoding the Cynomolgus SIGLEC6 (XP_005590181.3) (Met1-Gly333) was expressed with a polyhistidine tag at the C-terminus.