Learn More
Sino Biological Recombinant Cynomolgus TIE-2 Protein (hFc Tag)
Supplier:  Sino Biological 90292C02H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus TEK (G7PS68) (Met1-Lys745) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3088