Learn More
Sino Biological Recombinant Cynomolgus TGF beta 1 Protein (Latent, His Tag)
Supplier:  Sino Biological 90294C08H500
Encompass_Preferred
A DNA sequence encoding the cynomolgus TGFB1 (XP_005589396.1) (Met1-Ser390) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6876