Learn More
Sino Biological Anti-TPO/Thrombopoietin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90004RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus Thrombopoietin / THPO (rCynomolgus Thrombopoietin / THPO; Catalog#90004-C08H; XP_005546625.1; Met 1-Gly 353). Thrombopoietin / THPO specific IgG was purified by Cynomolgus Thrombopoietin / THPO affinity chromatography.
Catalog No. 50-241-2041