Learn More
Sino Biological Recombinant Cynomolgus TIGIT Protein (ECD, hFc Tag) 1
Supplier:  Sino Biological 90890C02H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus TIGIT (XP_015300911.1) (Met1-Pro209) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6917