Learn More
Sino Biological Recombinant Cynomolgus TIM-3/HAVCR2 Protein (hFc Tag) 1
Supplier:  Sino Biological 90312C02H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus TIM3 (EHH54703.1) (Met1-Arg201) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7156