Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Cynomolgus (Macaca fascicularis) TNF-alpha Protein, His Tag, Recombinant (HEK293), 100 μg

Catalog No. NC3761936
Encompass_Preferred

Recombinant cynomolgus (Macaca fascicularis) tumor necrosis factor alpha (TNF-alpha) produced in human HEK293 cells. The protein is His-tagged and MALS-verified for oligomeric state, supplied in a 100 μg format, and intended for immunoassay development, antibody screening, and functional studies.

  • Expression system: human 293 (HEK293) cells.
  • Amino acid range: Val 77-Leu 233.
  • Accession number: P79337.
  • Tag: His tag.
  • Quality: MALS verified for oligomeric state.
  • Available size: 100 μg.
  • Applications: immunoassay development, antibody screening, functional studies.

Catalog No. NC3761936 Supplier ACROBiosystems Supplier No. TNAC52H4
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant cynomolgus (Macaca fascicularis) tumor necrosis factor alpha (TNF-alpha) produced in human HEK293 cells. The protein is His-tagged and MALS-verified for oligomeric state, supplied in a 100 μg format, and intended for immunoassay development, antibody screening, and functional studies.

  • Expression system: human 293 (HEK293) cells.
  • Amino acid range: Val 77-Leu 233.
  • Accession number: P79337.
  • Tag: His tag.
  • Quality: MALS verified for oligomeric state.
  • Available size: 100 μg.
  • Applications: immunoassay development, antibody screening, functional studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.