Learn More
Sino Biological Recombinant Cynomolgus TACI Protein (His Tag)
Supplier:  Sino Biological 90976C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus TNFRSF13B (XP_005583052.1) (Ser2-Leu160) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2639