Learn More
Sino Biological Recombinant Cynomolgus CD30/TNFRSF8 Protein (His Tag)
Supplier:  Sino Biological 90899C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus TNFRSF8 (XP_015298065.1) (Met1-Gly194) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2660