Learn More
Sino Biological Cynomolgus TNFSF11 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90301C01H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus TNFSF11 (G7PW41) (Gly136-Asp317) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-2073