Learn More
Sino Biological Anti-CD30 Ligand/TNFSF8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90091T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus CD153 / CD30L / TNFSF8 (Catalog#90091-C07H; G7P2F1; Gln63-Asp234). CD153 / CD30L / TNFSF8 specific IgG was purified by Cynomolgus CD153 / CD30L / TNFSF8 affinity chromatography.
Catalog No. 50-255-2720