Learn More
Sino Biological Cynomolgus TNFSF9 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90848C01H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus TNFSF9 (XP_015296398.1) (Arg68-Glu251) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-3148