Learn More
Sino Biological Cynomolgus TP53 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 90001MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus p53 / TP53 (rC p53 / TP53; Catalog#90001-CNAE; E3U906; Met 1-Asp 393). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3920