Learn More
ACROBiosystems Cynomolgus TRAIL R2 / DR5 / TNFRSF10B Protein, His Tag
Supplier:  ACROBiosystems TR2C52H71MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus TRAIL R2, His Tag (TR2-C52H7) is expressed from human 293 cells (HEK293). It contains AA Ile 58 - Ser 212 (Accession # A0A2K5TXK0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.1 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Cynomolgus TRAIL R2, His Tag (Cat. No. TR2-C52H7) at 5 μg/mL (100 μL/well) can bind Human TRAIL, Mouse IgG2a Fc Tag (Cat. No. TRL-H5259) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-205-5255