Learn More
ACROBiosystems Biotinylated Cynomolgus Transferrin R, His Tag, primary amine labeling (TFR-C8249) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # G8F602-1).
Catalog No. 502230818
Encompass_Preferred
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia & lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.
This item is not returnable. View return policy
Catalog No. 50-223-0818 Supplier ACROBiosystems Supplier No. TFRC824925UG
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia & lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.