Learn More
ACROBiosystems Cynomolgus TREM2 Protein, His Tag
Catalog No. 502054903
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus TREM2, His Tag (TR2-C52H3) is expressed from human 293 cells (HEK293). It contains AA His 19 - Ser 174 (Accession # A0A2K5UY39-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4903 Supplier ACROBiosystems Supplier No. TR2C52H3100UG
Recombinant Protein;100UG;Cynomolgus TREM2, His Tag (TR2-C52H3) is expressed from human 293 cells (HEK293). It contains AA His 19 - Ser 174 (Accession # A0A2K5UY39-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.