Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Cynomolgus TROP-2 (TACSTD2) recombinant extracellular domain, C-terminal His-tag, lyophilized, 1 mg

Catalog No. NC2880829
Encompass_Preferred

Cynomolgus TROP-2 (TACSTD2) recombinant extracellular domain produced in HEK293 cells and supplied as a lyophilized C-terminal His-tagged protein. The protein is verified by SEC-MALS and intended for use in antibody binding assays and other in vitro applications.

  • Recombinant extracellular domain of TROP-2 (AA Gln 31-Thr 274)
  • Expressed in human HEK293 cells
  • C-terminal polyhistidine (His) tag
  • Purity greater than 90% by SDS-PAGE and SEC-MALS
  • Calculated molecular weight 29.4 kDa; appears 40-53 kDa on SDS-PAGE due to glycosylation
  • Verified by SEC-MALS, SDS-PAGE, ELISA, and SPR
  • Endotoxin less than 1.0 EU per μg (LAL/rFC)
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4, with trehalose
  • Available in 100 μg and 1 mg sizes
  • Store lyophilized at -20°C or lower; avoid repeated freeze-thaw

Catalog No. NC2880829 Supplier ACROBiosystems Supplier No. TR2C52H51MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cynomolgus TROP-2 (TACSTD2) recombinant extracellular domain produced in HEK293 cells and supplied as a lyophilized C-terminal His-tagged protein. The protein is verified by SEC-MALS and intended for use in antibody binding assays and other in vitro applications.

  • Recombinant extracellular domain of TROP-2 (AA Gln 31-Thr 274)
  • Expressed in human HEK293 cells
  • C-terminal polyhistidine (His) tag
  • Purity greater than 90% by SDS-PAGE and SEC-MALS
  • Calculated molecular weight 29.4 kDa; appears 40-53 kDa on SDS-PAGE due to glycosylation
  • Verified by SEC-MALS, SDS-PAGE, ELISA, and SPR
  • Endotoxin less than 1.0 EU per μg (LAL/rFC)
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4, with trehalose
  • Available in 100 μg and 1 mg sizes
  • Store lyophilized at -20°C or lower; avoid repeated freeze-thaw

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.