Learn More
Sino Biological Recombinant Cynomolgus TSLP Protein (His Tag)
Supplier:  Sino Biological 90911C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus TSLP ( XP_005557555.1, with mutation Arg 127 Ala, Arg 130 Ser) (Met1-Gln159) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6778