Learn More
Sino Biological Recombinant Cynomolgus TSPAN8 Protein (AVI & His Tag), Avi-tag Biotinylated
Catalog No. 502992960
Encompass_Preferred
A DNA sequence encoding the Cynomolgus TSPAN8 (G7PI18-1) (Lys110-Ser205) was expressed with an N-terminal AVI tagged polyhistidine tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2960 Supplier Sino Biological Supplier No. 91064C44HB100
A DNA sequence encoding the Cynomolgus TSPAN8 (G7PI18-1) (Lys110-Ser205) was expressed with an N-terminal AVI tagged polyhistidine tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.