Learn More
Sino Biological Cynomolgus Vsir Protein 500ug (Sino Biological)
Supplier:  Sino Biological 90844C02H500
Encompass_Preferred
A DNA sequence encoding the cynomolgus B7H5 (XP_005565644.1) (Met1-Ala194) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2027