Learn More
Cayman Chemical CyntrnIlIprole 50mg
SDP
Catalog No. 502941243
Encompass_Preferred
An anthranilic diamide insecticide; an allosteric RyR activator; 300- to 500-fold selective for insect over mouse RyR1, greater than 2,000-fold selective for insect over rat RyR2, and inactive in human IMR32 cells expressing RyR2 and RyR3; active against the diamondback moth (PP50 = 0.1 ppm); induces mortality of the green peach aphid, cotton melon aphid, and white fly (EC50s = 1.1, 0.4, and 5.8 ppm, respectively)
Catalog No. 50-294-1243 Supplier Cayman Chemical Supplier No. 2730950 MG
An anthranilic diamide insecticide; an allosteric RyR activator; 300- to 500-fold selective for insect over mouse RyR1, greater than 2,000-fold selective for insect over rat RyR2, and inactive in human IMR32 cells expressing RyR2 and RyR3; active against the diamondback moth (PP50 = 0.1 ppm); induces mortality of the green peach aphid, cotton melon aphid, and white fly (EC50s = 1.1, 0.4, and 5.8 ppm, respectively)