Learn More
Aviva Systems Biology Corp CYP11A1 Antibody - middle region (ARP51905_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51905_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP11A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP11A1. Alternate names: CYP11A, P450SCC, CYPXIA1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-60642