Learn More
Aviva Systems Biology Corp CYP11B1 Antibody - C-terminal region (ARP60140_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60140_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP11B1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CYP11B1. Alternate names: CPN1, CYP11B, DKFZp686B05283, FHI, FLJ36771, P450C11ApplicationIHC, WB. Species reactivity: Human, Pig, Sheep
Catalog No. 50-286-957