Learn More
Aviva Systems Biology Corp CYP11B2 Antibody - middle region (ARP41750_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41750_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP11B2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP11B2. Alternate names: ALDOS, CPN2, CYP11B, CYP11BL, P-450C18, P450C18, P450aldoApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-57980