Learn More
Biomatik Corporation Cyp19A1 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07021
Encompass_Preferred
Cyp19A1 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to CYP19A1. The Cyp19A1 Recombinant Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Cyp19A1. Applications: ELISA, WB, IHC, IF. Target Synonyms: CYP19A1. Accession Number: P11511. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5416