Learn More
Aviva Systems Biology Corp CYP1A1 Antibody - middle region (ARP41405_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41405_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP1A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP1A1. Alternate names: AHH, AHRR, CP11, CYP1, P1-450, P450-C, P450DXApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57836