Learn More
Cayman Chemical CYP1Acod Monclonal ntIbo 500uL
SDP
Catalog No. 502965312
Encompass_Preferred
An inhibitor of H-PGDS (IC50 32 nM for the human enzyme) selective for H-PGDS over L-PGDS mPGES-1 COX-1 COX-2 and 5-LO (IC50s 10000 nM for all) inhibits PGD2 production in vivo in rat spleen at 1 mg/kg or 10 mg/kg completely inhibits allergen-induced airway hyperresponsiveness in an ovine model of asthma
Catalog No. 50-296-5312 Supplier Cayman Chemical Supplier No. 173125500 UL
An inhibitor of H-PGDS (IC50 32 nM for the human enzyme) selective for H-PGDS over L-PGDS mPGES-1 COX-1 COX-2 and 5-LO (IC50s 10000 nM for all) inhibits PGD2 production in vivo in rat spleen at 1 mg/kg or 10 mg/kg completely inhibits allergen-induced airway hyperresponsiveness in an ovine model of asthma