Learn More
Cayman Chemical CYP1AfIsh PolycLNal ntIb 500uL
SDP
Catalog No. 502967397
Encompass_Preferred
An ATP-competitive inhibitor of Hsp90 that causes degradation of client proteins resulting in cell cycle arrest or apoptosis orally bioavailable and shows efficacy in tumor regression in gastric carcinoma and melanoma xenografts in mice
Catalog No. 50-296-7397 Supplier Cayman Chemical Supplier No. 173135500 UL
An ATP-competitive inhibitor of Hsp90 that causes degradation of client proteins resulting in cell cycle arrest or apoptosis orally bioavailable and shows efficacy in tumor regression in gastric carcinoma and melanoma xenografts in mice