Learn More
Aviva Systems Biology Corp CYP1B1 Antibody - middle region (ARP51761_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51761_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP1B1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CYP1B1. Alternate names: CP1B, GLC3A, P4501B1, CYPIB1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60607