Learn More
Aviva Systems Biology Corp CYP21A2 Antibody - C-terminal region (ARP60144_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60144_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP21A2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CYP21A2. Alternate names: CA21H, CAH1, CPS1, CYP21, CYP21B, MGC150536, MGC150537, P450c21BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Pig, Sheep
Catalog No. 50-286-959