Learn More
Aviva Systems Biology Corp CYP24A1 Antibody - C-terminal region (ARP60146_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60146_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP24A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CYP24A1. Alternate names: CP24, CYP24, MGC126273, MGC126274, P450-CC24, HCAIApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-960