Learn More
ABclonal Technology CYP24A1 Rabbit mAb
SDP
Catalog No. 5000006848
Encompass_Preferred
This gene encodes a member of the cytochrome P450 superfamily of enzymes The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol steroids and other lipids This mitochondrial protein initiates the degradation of 1 25-dihydroxyvitamin D3 the physiologically active form of vitamin D3 by hydroxylation of the side chain In regulating the level of vitamin D3 this enzyme plays a role in calcium homeostasis and the vitamin D endocrine system Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-06848 Supplier ABclonal Technology Supplier No. A22615100UL
This gene encodes a member of the cytochrome P450 superfamily of enzymes The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol steroids and other lipids This mitochondrial protein initiates the degradation of 1 25-dihydroxyvitamin D3 the physiologically active form of vitamin D3 by hydroxylation of the side chain In regulating the level of vitamin D3 this enzyme plays a role in calcium homeostasis and the vitamin D endocrine system Alternatively spliced transcript variants encoding different isoforms have been found for this gene