Learn More
Aviva Systems Biology Corp CYP26B1 Antibody - middle region (ARP57352_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57352_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP26B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP26B1. Alternate names: CYP26A2, DKFZp686G0638, MGC129613, P450RAI-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53974