Learn More
Aviva Systems Biology Corp CYP27A1 Antibody - middle region (ARP51773_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51773_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP27A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP27A1. Alternate names: CP27, CTX, CYP27ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60611