Learn More
Aviva Systems Biology Corp CYP2A13 Antibody - C-terminal region (ARP41803_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41803_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2A13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CYP2A13. Alternate names: CPAD, CYP2A, CYPIIA13ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-57999