Learn More
Aviva Systems Biology Corp CYP2A6 Antibody - C-terminal region (ARP60126_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60126_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2A6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CYP2A6. Alternate names: CPA6, CYP2A, CYP2A3, P450C2A, P450PB, CYPIIA6ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-286-952