Learn More
Aviva Systems Biology Corp CYP2A7 Antibody - N-terminal region (ARP41799_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41799_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2A7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CYP2A7. Alternate names: CPA7, CPAD, CYP2A, CYPIIA7, P450-IIA4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-957