Learn More
Aviva Systems Biology Corp CYP2B6 Antibody - middle region (ARP41805_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2B6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP2B6. Alternate names: CPB6, CYP2B, CYPIIB6, IIB1, P450, CYP2B7, CYP2B7PApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58000