Learn More
Cayman Chemical CYP2B6 Induction Reporter Assay Kit, 1 ea
SDP
Catalog No. 502893367
Encompass_Preferred
Early screening of a drug candidate’s potential to induce or inhibit CYP2B6 is a critical step in the drug development process.; Cayman's CYP2B6 Induction Reporter Assay Kit is a novel method to evaluate human Constitutive Androstane Receptor 3 (hCAR3)-mediated CYP2B6 induction for the discovery of hepatic CYP inducers.; Cayman's CYP2B6 Induction Reporter Assay Kit consists of a 96-well plate coated with a transfection complex containing a Secreted Alkaline Phosphatase (SEAP) reporter regulated by the human CYP2B6 gene promoter.; Binding of inducer-activated receptor and transcription factors to the CYP2B6 promoter initiates a signal resulting in expression of SEAP, which is secreted into the cell culture medium.; Aliquots of medium are removed at time intervals beginning at about 24 hours and SEAP activity is measured simply by adding a luminescence-based alkaline phosphatase substrate provided in the kit.
Catalog No. 50-289-3367 Supplier Cayman Chemical Supplier No. 6006801 EA
Early screening of a drug candidate’s potential to induce or inhibit CYP2B6 is a critical step in the drug development process.; Cayman's CYP2B6 Induction Reporter Assay Kit is a novel method to evaluate human Constitutive Androstane Receptor 3 (hCAR3)-mediated CYP2B6 induction for the discovery of hepatic CYP inducers.; Cayman's CYP2B6 Induction Reporter Assay Kit consists of a 96-well plate coated with a transfection complex containing a Secreted Alkaline Phosphatase (SEAP) reporter regulated by the human CYP2B6 gene promoter.; Binding of inducer-activated receptor and transcription factors to the CYP2B6 promoter initiates a signal resulting in expression of SEAP, which is secreted into the cell culture medium.; Aliquots of medium are removed at time intervals beginning at about 24 hours and SEAP activity is measured simply by adding a luminescence-based alkaline phosphatase substrate provided in the kit.