Learn More
Aviva Systems Biology Corp CYP2C18 Antibody - N-terminal region (ARP41810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2C18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CYP2C18. Alternate names: CPCI, CYP2C, CYP2C17, DKFZp686I24235, P450-6B/29C, P450IIC17ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58002