Learn More
Aviva Systems Biology Corp CYP2C19 Antibody - middle region (ARP41807_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41807_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2C19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP2C19. Alternate names: CPCJ, CYP 2C, CYP2C, P450C2C, P450IIC19ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53315