Learn More
Aviva Systems Biology Corp CYP2C9 Antibody - C-terminal region (ARP41809_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41809_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2C9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CYP2C9. Alternate names: CPC9, CYP2C, CYP2C10, MGC149605, MGC88320, P450 MP-4, P450 PB-1, P450IIC9, CYPIIC9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58001