Learn More
Aviva Systems Biology Corp CYP2D6 Antibody - N-terminal region (ARP41675_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41675_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2D6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CYP2D6. Alternate names: RP4-669P10.2, CPD6, CYP2D, CYP2D@, CYP2DL1, MGC120389, MGC120390, P450-DB1, P450C2D, CYPIID6, P450DB1, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57944