Learn More
Aviva Systems Biology Corp CYP2D6 Antibody - middle region (ARP41676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2D6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP2D6. Alternate names: CPD6, CYP2D, CYP2D@, CYP2DL1, MGC120389, MGC120390, P450-DB1, P450C2D, CYPIID6, P450DB1, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-57945