Learn More
Aviva Systems Biology Corp CYP2E1 Antibody - C-terminal region (ARP41424_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41424_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2E1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CYP2E1. Alternate names: CPE1, CYP2E, P450-J, P450C2EApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-57843