Learn More
Medchemexpress LLC CYP2E1-IN-1 | 38205-55-9 | MFCD09029046 | 100.0% | 141.19 g/mol | C6H7NOS | 250 MG
SDP
Catalog No. 5000208689
Encompass_Preferred
CYP2E1-IN-1 (CAS 38205-55-9) is a small-molecule cytochrome P450 2E1 inhibitor used for biochemical and pharmacological research. The compound is a low-molecular-weight solid (C6H7NOS, 141.19 g/mol) typically supplied at high purity for laboratory studies.
  • Inhibits cytochrome P450 2E1 activity
  • Suitable for in vitro and in vivo research
  • Available in multiple pack sizes, including 250 mg
  • Soluble in DMSO for assay preparation
Catalog No. 50-002-08689 Supplier Medchemexpress LLC Supplier No. 250MGHYW072025
CYP2E1-IN-1 (CAS 38205-55-9) is a small-molecule cytochrome P450 2E1 inhibitor used for biochemical and pharmacological research. The compound is a low-molecular-weight solid (C6H7NOS, 141.19 g/mol) typically supplied at high purity for laboratory studies.
  • Inhibits cytochrome P450 2E1 activity
  • Suitable for in vitro and in vivo research
  • Available in multiple pack sizes, including 250 mg
  • Soluble in DMSO for assay preparation