Learn More
Aviva Systems Biology Corp Cyp2f4 Antibody - middle region (ARP60130_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60130_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cyp2f4. It was validated on Western Blot. . Alternate names: CYP4502F4, Cyp2f1|Cyp2f2, Cyp2f1, Cyp2f2ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-953