Learn More
Aviva Systems Biology Corp CYP2R1 Antibody - middle region (ARP49823_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49823_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP2R1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CYP2R1. Alternate names: MGC4663ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Zebrafish
Catalog No. 50-285-455