Learn More
Medchemexpress LLC CYP3A4 Human His 10ug
SDP
Catalog No. 5000178479
Encompass_Preferred
The CYP3A4 protein encoded by the CYP3A4 gene is a key enzyme in drug metabolism and the synthesis of cholesterol steroids and lipids It localizes to the endoplasmic reticulum and is induced by glucocorticoids and certain drugs CYP3A4 Protein Human (His) is the recombinant human-derived CYP3A4 protein expressed by E coli with N-10 His labeled tag
Catalog No. 50-001-78479 Supplier Medchemexpress LLC Supplier No. HYP7421010UG
The CYP3A4 protein encoded by the CYP3A4 gene is a key enzyme in drug metabolism and the synthesis of cholesterol steroids and lipids It localizes to the endoplasmic reticulum and is induced by glucocorticoids and certain drugs CYP3A4 Protein Human (His) is the recombinant human-derived CYP3A4 protein expressed by E coli with N-10 His labeled tag